Please login to your online account to display your discounted pricing

p-Dioxane, PHOTREX™ Reagent, J.T.Baker™

High purity solvents characterized for use in UV, visible and IR spectrophotometry

$533.11 - $1,068.91

Chemical Identifiers

Chemical Name or Material p-Dioxane
CAS 123-91-1
Purity Grade PHOTREX™ Reagent
Molecular Formula C4H8O2
Formula Weight 88.11
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02003529

 
avantor performance
919602
Narrow Mouth Amber Glass Bottle 1L Each for $533.11

02003531

 
avantor performance
919603
Narrow Mouth Amber Glass Bottle 4L Each for $1,068.91
Description & Specifications

Specifications

Formula Weight 88.11
Chemical Name or Material p-Dioxane
Packaging Narrow Mouth Amber Glass Bottle
Purity Grade PHOTREX™ Reagent
Molecular Formula C4H8O2
CAS 123-91-1
Flash Point 12.00°C
Specific Gravity 1L = 1.03kg
Quantity 1L

    For Spectrophotometry and Liquid Scintillation Counting