Please login to your online account to display your discounted pricing

α,p-Dimethylstyrene 95%, stabilized, 95%, Acros Organics


Quantity 100g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC386291000

 View more versions of this product

Catalog No. AC386291000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥94%
Physical Form Liquid
Formula Weight 132.21
Quantity 100g
Color Clear colorless to light yellow
Density 0.9040g/mL
CAS Min % 94.0
CAS Max % 100.0
Chemical Name or Material α, p-Dimethylstyrene
Linear Formula CH3C6H4C(CH3)=CH2
MDL Number MFCD00036510
CAS 1195-32-0
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.5300 to 1.5390 (20°C, 589nm)
Assay Percent Range 95%
Molecular Formula C10H12
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

Specifications

Chemical Name or Material α, p-Dimethylstyrene
CAS 1195-32-0
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C10H12
Linear Formula CH3C6H4C(CH3)=CH2
Formula Weight 132.21
MDL Number MFCD00036510
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents