Please login to your online account to display your discounted pricing

α,p-Dimethylstyrene 95%, stabilized, 95%, Acros Organics


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC386290250

 View more versions of this product

Catalog No. AC386290250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥94%
Formula Weight 132.21
Physical Form Liquid
Quantity 25g
Color Clear colorless to light yellow
Density 0.9040g/mL
CAS Max % 100.0
CAS Min % 94.0
Packaging Glass bottle
Linear Formula CH3C6H4C(CH3)=CH2
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
MDL Number MFCD00036510
Assay Percent Range 95%
Infrared Spectrum Authentic
Chemical Name or Material α, p-Dimethylstyrene
Refractive Index 1.5300 to 1.5390 (20°C, 589nm)
Molecular Formula C10H12
CAS 1195-32-0

Specifications

Chemical Name or Material α, p-Dimethylstyrene
CAS 1195-32-0
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C10H12
Linear Formula CH3C6H4C(CH3)=CH2
Formula Weight 132.21
MDL Number MFCD00036510
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents