Please login to your online account to display your discounted pricing

α,p-Dimethylstyrene 95%, stabilized, 95%, Acros Organics

Chemical Identifiers

Chemical Name or Material α, p-Dimethylstyrene
CAS 1195-32-0
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C10H12
Linear Formula CH3C6H4C(CH3)=CH2
Formula Weight 132.21
MDL Number MFCD00036510
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC386290050

  SDS
thermo fisher scientific
AC386290050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC386290250

  SDS
thermo fisher scientific
AC386290250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC386291000

  SDS
thermo fisher scientific
AC386291000
Glass bottle 100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥94%
Physical Form Liquid
Formula Weight 132.21
Quantity 5g
Color Clear colorless to light yellow
Density 0.9040g/mL
CAS Min % 94.0
CAS Max % 100.0
Linear Formula CH3C6H4C(CH3)=CH2
Chemical Name or Material α, p-Dimethylstyrene
Packaging Glass bottle
Refractive Index 1.5300 to 1.5390 (20°C, 589nm)
Assay Percent Range 95%
MDL Number MFCD00036510
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Molecular Formula C10H12
CAS 1195-32-0
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.