Please login to your online account to display your discounted pricing

p-Dimethylaminobenzaldehyde (Certified ACS), Fisher Chemical


Quantity 100g
Packaging Glass Bottle

Manufacturer: fisher scientific company  D71100

 View more versions of this product

Catalog No. D71-100

  • / Each
  • / Case of 6 Each
 

Description & Specifications

Chemical Identifiers

Purity Grade Certified ACS Grade
Physical Form Powder/Solid
Formula Weight 149.19
Boiling Point 176°C
Melting Point 70°C
Quantity 100g
Density 5.15 (Air=1.0)
Flash Point 164°C
Molecular Formula C9H11NO
Alpha Vector Dimethylaminobenzaldehyde
Color Light Yellow
Packaging Glass Bottle
Solubility Pass Test (in HCl)
Solubility in Alcohol Pass Test
ChemAlert Storage Symbol Gray
Synonym 4-Dimethylaminobenzenecarbonal
Identification Pass Test
Ignition Residue 0.1% max.
Chemical Name or Material p-Dimethylaminobenzaldehyde
Linear Formula (CH3)2NC6H4CHO
MDL Number MFCD00003381
Health Hazard 2 WARNING!
Health Hazard 3 Emergency Overview
Harmful if swallowed. Harmful to aquatic organisms, may cause long-term adverse effects in the aquatic environment. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Get medical attention immediately if symptoms occur. Get medical attention immediately if symptoms occur. Do not induce vomiting.

NFPA

Health:2
Flammability:1
Instability:1
CAS 100-10-7

Specifications

Chemical Name or Material p-Dimethylaminobenzaldehyde
CAS 100-10-7
Purity Grade Certified ACS Grade
Physical Form Powder/Solid
Molecular Formula C9H11NO
Linear Formula (CH3)2NC6H4CHO
Formula Weight 149.19
MDL Number MFCD00003381
Synonym 4-Dimethylaminobenzenecarbonal
Accessories
 

Emergency Overview
Harmful if swallowed. Harmful to aquatic organisms, may cause long-term adverse effects in the aquatic environment. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Get medical attention immediately if symptoms occur. Get medical attention immediately if symptoms occur. Do not induce vomiting.

NFPA

Health:2
Flammability:1
Instability:1

WARNING!

SDS & Safety Documents