Please login to your online account to display your discounted pricing

p-Dimethylaminobenzaldehyde, ACS Grade, LabChem™

$267.66 - $267.66

Chemical Identifiers

Chemical Name or Material p-Dimethylaminobenzaldehyde
CAS 100-10-7
CAS Max % 100
Purity Grade ACS Grade
Linear Formula C9H11NO
Formula Weight 149.19
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

LC135899

 
lab chem
LC135899
Amber Glass 100g Each for $267.66
Description & Specifications

Specifications

Formula Weight 149.19
Chemical Name or Material p-Dimethylaminobenzaldehyde
Packaging Amber Glass
Solubility Soluble in water
Purity Grade ACS Grade
Physical Form Powder
Storage Information Room Temperature
Linear Formula C9H11NO
CAS 100-10-7
Health Hazard 1 Warning
CAS Max % 100
Health Hazard 2 GHS H Statement
Causes serious eye irritation.
Identification Passes Test
Health Hazard 3 GHS P Statement
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Decomposition Information Nitrogen oxides; Carbon monoxide; Carbon dioxide
Color White to pale yellow
Melting Point 74°C
Quantity 100g
Safety and Handling

Warning

GHS P Statement
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
Causes serious eye irritation.