missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

p-Cresol, +99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 405740050

 View more versions of this product

Catalog No. AC405740050


 

Specifications Description & Specifications

Specifications

≥99.0%
108.14
Colorless to light yellow, may darken on exposure to light
p-Cresol
4-Hydroxytoluene
99+%
CH3C6H4OH
C7H8O
Glass bottle
Crystalline Solid or Liquid
5g
Pure
0.05% max.
Authentic
106-44-5
99+%
MFCD00002376

Chemical Identifiers

106-44-5
C7H8O
108.14
4-Hydroxytoluene
≥99.0%
99+%
CH3C6H4OH
MFCD00002376
p-Cresol, 99+%
Pure

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes serious eye damage.
Fatal in contact with skin.
Causes severe skin burns and eye damage.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Remove/Take off immediately all contaminated clothing.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 203-398-6

RTECSNumber : GO6475000

TSCA : TSCA