Please login to your online account to display your discounted pricing

p-Chlorovalerophenone, 98%, Acros Organics


Quantity 100g

Manufacturer: thermo fisher scientific  240941000

 View more versions of this product

Catalog No. AC240941000

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Crystalline Low Melting Solid
Formula Weight 196.67
Quantity 100g
Color White to yellow
EINECS Number 246-566-4
Melting Point 29° to 33°C
Synonym 1-(4-Chlorophenyl)-1-pentanone
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Name Note 98%
CAS 25017-08-7
Linear Formula ClC6H5CO(CH2)3CH3
Assay Percent Range 98%
Chemical Name or Material p-Chlorovalerophenone
MDL Number MFCD00018716
Molecular Formula C11H13ClO
Infrared Spectrum Authentic

Specifications

Chemical Name or Material p-Chlorovalerophenone
Name Note 98%
CAS 25017-08-7
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C11H13ClO
Linear Formula ClC6H5CO(CH2)3CH3
Formula Weight 196.67
MDL Number MFCD00018716
Synonym 1-(4-Chlorophenyl)-1-pentanone
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

SDS & Safety Documents