missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

p-Chlorovalerophenone, 98%, Acros Organics

Manufacturer:  Thermo Fisher Scientific 240940050

 View more versions of this product

Catalog No. AC240940050

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

p-Chlorovalerophenone
29° to 33°C
98%
Authentic
≥97.5%
C11H13ClO
196.67
1-(4-Chlorophenyl)-1-pentanone
White to yellow
5g
25017-08-7
Crystalline Low Melting Solid
98%
ClC6H5CO(CH2)3CH3
MFCD00018716

Chemical Identifiers

25017-08-7
C11H13ClO
196.67
1-(4-Chlorophenyl)-1-pentanone
≥97.5%
98%
ClC6H5CO(CH2)3CH3
MFCD00018716
p-Chlorovalerophenone, 98%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 246-566-4