missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

p-Chlorovalerophenone, 98%, Acros Organics

$299.05 - $299.05

Chemical Identifiers

CAS 25017-08-7
Assay Percent Range 98%
Molecular Formula C11H13ClO
Linear Formula ClC6H5CO(CH2)3CH3
Formula Weight 196.67
MDL Number MFCD00018716
Synonym 1-(4-Chlorophenyl)-1-pentanone
Chemical Name or Material p-Chlorovalerophenone, 98%
Assay ≥97.5%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC240940050 SDS Thermo Fisher Scientific
240940050
5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC240941000 SDS Thermo Fisher Scientific
240941000
100g Each for $299.05
Specifications Description & Specifications

Chemical Identifiers

25017-08-7
C11H13ClO
196.67
1-(4-Chlorophenyl)-1-pentanone
≥97.5%
98%
ClC6H5CO(CH2)3CH3
MFCD00018716
p-Chlorovalerophenone, 98%

Specifications

White to yellow
5g
Authentic
98%
ClC6H5CO(CH2)3CH3
MFCD00018716
p-Chlorovalerophenone, 98%
29° to 33°C
25017-08-7
Crystalline Low Melting Solid
C11H13ClO
196.67
1-(4-Chlorophenyl)-1-pentanone
≥97.5%

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 246-566-4