missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

p-Chloranil, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1349536

 View more versions of this product

Catalog No. AAA1349536



Description

Description

p-Chloranil is used as a dye intermediate, oxidizing agent, vulcanizing agent and dehydrogenation reagent. It is also used to make chloranil electrodes for pH measurements. Further, it serves as a hydrogen acceptor and used for the aromatization reactions such as conversion of cyclohexadienes to the benzene derivatives. It is used to test the secondary amine. In addition to this, it is employed as a precursor to prepare diaziquone, which is used as a cancer chemotherapeutic agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
p-Chloranil is used as a dye intermediate, oxidizing agent, vulcanizing agent and dehydrogenation reagent. It is also used to make chloranil electrodes for pH measurements. Further, it serves as a hydrogen acceptor and used for the aromatization reactions such as conversion of cyclohexadienes to the benzene derivatives. It is used to test the secondary amine. In addition to this, it is employed as a precursor to prepare diaziquone, which is used as a cancer chemotherapeutic agent.

Solubility
Soluble in ether and acetone. Slightly soluble in chloroform, dimethylformamide, solvent naphtha, benzene, carbon disulfide and carbon tetrachloride. Insoluble in water, cold petroleum ether, methanol, dibutyl phthalate and cold alcohol.

Notes
Incompatible with strong oxidizing agents and strong bases.
Specifications

Specifications

p-Chloranil
∼290°C (sublimation)
C6Cl4O2
500 g
393006
chloranil, p-chloranil, tetrachloro-p-benzoquinone, tetrachloro-1,4-benzoquinone, tetrachlorobenzoquinone, spergon, coversan, reranil, vulklor, 2,3,5,6-tetrachloro-1,4-benzoquinone
UGNWTBMOAKPKBL-UHFFFAOYSA-N
2,3,5,6-tetrachlorocyclohexa-2,5-diene-1,4-dione
8371
245.88
118-75-2
1.97
MFCD00001594
UN3077
14,2078
Soluble in ether and acetone. Slightly soluble in chloroform,dimethylformamide,solvent naphtha,benzene,carbon disulfide and carbon tetrachloride. Insoluble in water,cold petroleum ether,methanol,dibutyl phthalate and cold alcohol.
ClC1=C(Cl)C(=O)C(Cl)=C(Cl)C1=O
245.86
CHEBI:36703
97%

Chemical Identifiers

118-75-2
245.86
UGNWTBMOAKPKBL-UHFFFAOYSA-N
8371
2,3,5,6-tetrachlorocyclohexa-2,5-diene-1,4-dione
C6Cl4O2
MFCD00001594
chloranil, p-chloranil, tetrachloro-p-benzoquinone, tetrachloro-1,4-benzoquinone, tetrachlorobenzoquinone, spergon, coversan, reranil, vulklor, 2,3,5,6-tetrachloro-1,4-benzoquinone
CHEBI:36703
ClC1=C(Cl)C(=O)C(Cl)=C(Cl)C1=O
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P311-P332+P313-P362-P501c

H315-H319-H331

DOTInformation : Hazard Class: 6.1; Packaging Group: III

EINECSNumber : 204-274-4

RTECSNumber : DK6825000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only