Please login to your online account to display your discounted pricing

P-Cadherin Mouse anti-Human, Clone: 12H6, eBioscience™

Mouse Monoclonal Antibody

$129.05 - $288.55

Specifications

Antigen P-Cadherin
Clone 12H6
Host Species Mouse
Species Reactivity Human
Applications Immunocytochemistry
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

50245616

 
life technologies
LS14987380
25μg Each for $129.05

50245617

 
life technologies
LS14987382
100μg Each for $288.55
Description & Specifications

Specifications

Antigen P-Cadherin
Clone 12H6
Host Species Mouse
Species Reactivity Human
Applications Immunocytochemistry
Applications Immunohistochemistry (Formalin/Paraffin)
Applications Western Blotting
Regulatory Status RUO
Conjugate Unlabeled
Format Purified
Storage Requirements Store at 2-8°C.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

The monoclonal antibody 12H6 recognizes human P-cadherin (placental-cadherin), a calcium dependent cell-cell adhesion protein. This 118kDa transmembrane protein was originally named due to its expression in mouse placental tissue. Unfortunately, it is not expressed in human placental tissue. The function of the molecule will depend on the associations with the specific member of the cytoplasmic catenin family (alpha, beta and gamma). The specific interaction defines the strength and signaling of the cell-cell interaction. Expression is less broad than E-cadherin; P-cadherin is restricted to the basal proliferative cell layer of stratified epithelia. During development expression is found in the proliferating tissues such as hair follicle keratinocytes and the growth regions in ductal mammary tissue. There is evidence that P-cadherin can be secreted from epithelial cells during the late stages of pregnancy. P-cadherin expression is associated with poor prognosis for breast cancer while reduced expression in melanomas and squamous cell carcinomas may also result in poor prognosis.