Please login to your online account to display your discounted pricing

Alfa Aesar™ p-Arsanilic acid, 98%

Chemical Identifiers

Chemical Name or Material p-Arsanilic acid
CAS 98-50-0
Molecular Formula C6H8AsNO3
Formula Weight 217.06
MDL Number MFCD00007819
UN Number UN3465
Beilstein 1102334
Merck Index 14,792
Synonym 4-Aminobenzenearsonic acid; 4-Aminophenylarsonic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2459522

  SDS
alfa aesar™
B2459522
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2459536

  SDS
alfa aesar™
B2459536
500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 217.06
Chemical Name or Material p-Arsanilic acid
TSCA TSCA
Assay 98%
Molecular Formula C6H8AsNO3
MDL Number MFCD00007819
CAS 98-50-0
UN Number UN3465
Beilstein 1102334
Merck Index 14,792
Synonym 4-Aminobenzenearsonic acid; 4-Aminophenylarsonic acid
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H331
Toxic if swallowed.
Toxic if inhaled.
Melting Point >300°C
Quantity 100g
Health Hazard 3 GHS P Statement
P261-P301+P310-P304+P340-P311-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOARSENIC COMPOUND, SOLID, N.O.S.
RTECS Number CF7875000
Safety and Handling

Danger

GHS P Statement
P261-P301+P310-P304+P340-P311-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H331
Toxic if swallowed.
Toxic if inhaled.