missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

p-Anisylchlorodiphenylmethane, 97%, Thermo Scientific™

$444.86 - $444.86

Chemical Identifiers

CAS 14470-28-1
Molecular Formula C20H17ClO
Molecular Weight (g/mol) 308.8
MDL Number MFCD00000814
InChI Key OBOHMJWDFPBPKD-UHFFFAOYSA-N
Synonym 1-chlorodiphenylmethyl-4-methoxybenzene, 4-methoxytriphenylchloromethane, 4-methoxytriphenylmethyl chloride, 4-methoxytrityl chloride, 4-methoxytritylchloride, 4-monomethoxytrityl chloride, chloro 4-methoxyphenyl methylene dibenzene, mmtrcl, p-anisylchlorodiphenylmethane, p-chlorodiphenylmethyl anisole
PubChem CID 84462
IUPAC Name 1-[chloro(diphenyl)methyl]-4-methoxybenzene
SMILES COC1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)Cl
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC156561000
SDS View Documents Promotion Details
Thermo Fisher Scientific
156561000
100g Glass bottle
Each for $444.86
 
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

14470-28-1
308.8
OBOHMJWDFPBPKD-UHFFFAOYSA-N
84462
COC1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)Cl
C20H17ClO
MFCD00000814
1-chlorodiphenylmethyl-4-methoxybenzene, 4-methoxytriphenylchloromethane, 4-methoxytriphenylmethyl chloride, 4-methoxytrityl chloride, 4-methoxytritylchloride, 4-monomethoxytrityl chloride, chloro 4-methoxyphenyl methylene dibenzene, mmtrcl, p-anisylchlorodiphenylmethane, p-chlorodiphenylmethyl anisole
1-[chloro(diphenyl)methyl]-4-methoxybenzene

Specifications

14470-28-1
96% min
CH3OC6H4C(C6H5)2Cl
Glass bottle
100g
OBOHMJWDFPBPKD-UHFFFAOYSA-N
1-[chloro(diphenyl)methyl]-4-methoxybenzene
84462
97%
121.0°C to 125.0°C
C20H17ClO
MFCD00000814
06, 713
1-chlorodiphenylmethyl-4-methoxybenzene, 4-methoxytriphenylchloromethane, 4-methoxytriphenylmethyl chloride, 4-methoxytrityl chloride, 4-methoxytritylchloride, 4-monomethoxytrityl chloride, chloro 4-methoxyphenyl methylene dibenzene, mmtrcl, p-anisylchlorodiphenylmethane, p-chlorodiphenylmethyl anisole
COC1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)Cl
308.8
308.8
p-Anisylchlorodiphenylmethane, 97%
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

GHS Signal Word: Warning

EINECSNumber : 238-463-8

SDS
Product Certifications

RUO – Research Use Only