missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

p-Anisoyl Chloride 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 104851000

 View more versions of this product

Catalog No. AC104851000


 

Specifications Description & Specifications

Specifications

p-Anisoyl chloride
100-07-2
Liquid After Melting
99%
CH3OC6H4COCl
170.6
10, 163
1.5800 to 1.5820 (20°C, 589nm)
Clear colorless to brown
99%
Authentic
≥98.5%
C8H7ClO2
Glass bottle
MFCD00000687
15, 665
4-Methoxybenzoyl chloride
100g

Chemical Identifiers

100-07-2
C8H7ClO2
170.6
4-Methoxybenzoyl chloride
≥98.5%
99%
CH3OC6H4COCl
MFCD00000687
p-Anisoyl chloride, 99%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 202-816-4

TSCA : TSCA