Learn More
p-Anisidine hydrochloride, 99%, Thermo Scientific Chemicals
$113.70 - $557.52
Chemical Identifiers
CAS | 20265-97-8 |
---|---|
Molecular Formula | C7H9NO·HCl |
Molecular Weight (g/mol) | 159.61 |
MDL Number | MFCD00036388 |
InChI Key | VQYJLACQFYZHCO-UHFFFAOYSA-N |
Synonym | p-anisidine hydrochloride, 4-methoxyaniline hydrochloride, unii-7c4r75n8vg, benzenamine, 4-methoxy-, hydrochloride, p-anisidine.hcl, p-anisidine hcl, p-anisidinium chloride, 4-anisidine hydrochloride, p-anisylamine hydrochloride, p-aminoanisole hydrochloride |
PubChem CID | 2734956 |
IUPAC Name | 4-methoxyaniline;hydrochloride |
SMILES | COC1=CC=C(C=C1)N.Cl |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AC401300050
|
Thermo Scientific Chemicals
401300050 |
5 g |
Each for $113.70
|
|
|||||
AC401300250
|
Thermo Scientific Chemicals
401300250 |
25 g |
Each for $257.86
|
|
|||||
AC401301000
|
Thermo Scientific Chemicals
401301000 |
100 g |
Each for $557.52
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
20265-97-8 | |
159.61 | |
VQYJLACQFYZHCO-UHFFFAOYSA-N | |
2734956 | |
COC1=CC=C(C=C1)N.Cl |
C7H9NO·HCl | |
MFCD00036388 | |
p-anisidine hydrochloride, 4-methoxyaniline hydrochloride, unii-7c4r75n8vg, benzenamine, 4-methoxy-, hydrochloride, p-anisidine.hcl, p-anisidine hcl, p-anisidinium chloride, 4-anisidine hydrochloride, p-anisylamine hydrochloride, p-aminoanisole hydrochloride | |
4-methoxyaniline;hydrochloride |
Specifications
20265-97-8 | |
Brown-Gray to White | |
99% | |
C7H9NO·HCl | |
MFCD00036388 | |
p-anisidine hydrochloride, 4-methoxyaniline hydrochloride, unii-7c4r75n8vg, benzenamine, 4-methoxy-, hydrochloride, p-anisidine.hcl, p-anisidine hcl, p-anisidinium chloride, 4-anisidine hydrochloride, p-anisylamine hydrochloride, p-aminoanisole hydrochloride | |
VQYJLACQFYZHCO-UHFFFAOYSA-N | |
4-methoxyaniline;hydrochloride | |
2734956 | |
99% | |
p-Anisidine hydrochloride, 99% |
216.0°C to 219.0°C | |
Authentic | |
Glass bottle | |
NH2C6H4OCH3·HCl | |
5 g | |
Solubility in water: soluble. Other solubilities: soluble in ethanol | |
COC1=CC=C(C=C1)N.Cl | |
159.61 | |
159.61 | |
Crystalline Powder |
Safety and Handling
GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
May cause an allergic skin reaction.<br/
GHS P Statement
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Remove/Take off immediately all contaminated clothing.
IF ON SKIN: Wash with plenty of soap a
GHS Signal Word: Danger
EINECSNumber : 243-657-0
RTECSNumber : BZ6600000
TSCA : TSCA
RUO – Research Use Only