Learn More
p-Anisidine, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 104832500
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
p-Anisidine | |
104-94-9,90-04-0 | |
Brown to Gray | |
240.0°C to 243.0°C | |
Authentic | |
Glass bottle | |
CH3OC6H4NH2 | |
250 g | |
1.06 | |
p-anisidine, 4-anisidine, 4-methoxybenzenamine, 4-aminoanisole, p-aminoanisole, para-anisidine, p-methoxyaniline, benzenamine, 4-methoxy, 4-methoxybenzeneamine, p-anisylamine | |
BHAAPTBBJKJZER-UHFFFAOYSA-N | |
4-methoxyaniline | |
7732 | |
123.15 | |
Crystalline Powder, Granules or Flakes |
99% | |
56.0°C to 59.0°C | |
1.0600g/mL | |
122°C | |
98.5% min. (GC) | |
C7H9NO | |
MFCD00007864 | |
13, 435 | |
14, 667 | |
Solubility in water: 21g/L (20°C). Other solubilities: freely soluble in methanol, ethanol and ether, soluble in acetone and benzene | |
COC1=CC=C(C=C1)N | |
123.15 | |
CHEBI:82388 | |
99% |
Chemical Identifiers
104-94-9 | |
123.15 | |
BHAAPTBBJKJZER-UHFFFAOYSA-N | |
7732 | |
4-methoxyaniline |
C7H9NO | |
MFCD00007864 | |
p-anisidine, 4-anisidine, 4-methoxybenzenamine, 4-aminoanisole, p-aminoanisole, para-anisidine, p-methoxyaniline, benzenamine, 4-methoxy, 4-methoxybenzeneamine, p-anisylamine | |
CHEBI:82388 | |
COC1=CC=C(C=C1)N |
Safety and Handling
GHS H Statement
May cause damage to organs through prolonged or repeated exposure.
May cause cancer.
Fatal if inhaled.
Fatal in contact with skin.
Fatal if swallowed.
Very toxic to aquatic life.
GHS P Statement
Rinse mouth.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
GHS Signal Word: Danger
EINECSNumber : 203-254-2
RTECSNumber : BZ5450000
TSCA : TSCA
RUO – Research Use Only