missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
p-Anisic acid, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 152370051
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
p-Anisic acid | |
100-09-4 | |
185°C | |
97.5% min. (GC) | |
C8H8O3 | |
CH3OC6H4CO2H | |
5kg | |
15,659 | |
Solubility in water: 0.3g/L (20 c). Other solubilities: freely soluble in alcohol,ether,chloroform and,ethyl acetate | |
COC1=CC=C(C=C1)C(O)=O | |
152.15 | |
CHEBI:40813 | |
98% |
98% | |
275.0°C | |
Authentic | |
Plastic drum | |
1.5710 to 1.5760 | |
MFCD00002542 | |
10,IV,346 | |
p-anisic acid, anisic acid, p-methoxybenzoic acid, draconic acid, 4-anisic acid, benzoic acid, 4-methoxy, anisic acid, para, 4-methoxybenzoate, para-anisic acid, anisic acid, p-isomer | |
ZEYHEAKUIGZSGI-UHFFFAOYSA-N | |
4-methoxybenzoic acid | |
7478 | |
152.15 |
Safety and Handling
EINECSNumber : 202-818-5
RTECSNumber : BZ4395000
TSCA : TSCA
RUO â Research Use Only