missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
p-Anisaldehyde dimethyl acetal, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 352291000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
p-Anisaldehyde dimethyl acetal | |
1.0680g/mL | |
>110°C | |
97.5% min. (GC) | |
C10H14O3 | |
MFCD00036507 | |
1.068 | |
Solubility in water: insoluble. Other solubilities: soluble in alcohol,soluble in common organic solvents | |
COC(OC)C1=CC=C(OC)C=C1 | |
182.22 | |
182.22 |
2186-92-7 | |
235.0°C | |
Authentic | |
Glass bottle | |
1.5045 to 1.5065 | |
100g | |
1-dimethoxymethyl-4-methoxybenzene, p-anisaldehyde dimethyl acetal, anisaldehyde dimethyl acetal, 4-methoxybenzaldehyde dimethyl acetal, p-dimethoxymethyl anisole, benzene, 1-dimethoxymethyl-4-methoxy, p-anisaldehydedimethylacetal, unii-xte77a5et4, anisicaldehyde dimethylacetal, p-anisaldehyde dimethylacetal | |
NNHYAHOTXLASEA-UHFFFAOYSA-N | |
1-(dimethoxymethyl)-4-methoxybenzene | |
75140 | |
98% |
Chemical Identifiers
2186-92-7 | |
182.22 | |
NNHYAHOTXLASEA-UHFFFAOYSA-N | |
75140 | |
COC(OC)C1=CC=C(OC)C=C1 |
C10H14O3 | |
MFCD00036507 | |
1-dimethoxymethyl-4-methoxybenzene, p-anisaldehyde dimethyl acetal, anisaldehyde dimethyl acetal, 4-methoxybenzaldehyde dimethyl acetal, p-dimethoxymethyl anisole, benzene, 1-dimethoxymethyl-4-methoxy, p-anisaldehydedimethylacetal, unii-xte77a5et4, anisicaldehyde dimethylacetal, p-anisaldehyde dimethylacetal | |
1-(dimethoxymethyl)-4-methoxybenzene |
Safety and Handling
EINECSNumber : 218-577-4
RUO â Research Use Only