Please login to your online account to display your discounted pricing

Ozagrel hydrochloride, Tocris

Selective thromboxane A2 synthetase inhibitor

$142.08 - $598.33

Chemical Identifiers

Chemical Name or Material Ozagrel hydrochloride
Assay Percent Range >98%
Molecular Formula C13H12N2O2·HCl
Formula Weight 264.71
Synonym (2E)-3-[4-(1H-Imidazol-1-ylmethyl)phenyl]-2-propenoic acid hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

151010

 
r & d systems
RDS151010
10mg Each for $142.08

151050

 
r & d systems
RDS151050
50mg Each for $598.33
Description & Specifications

Specifications

Formula Weight 264.71
Quantity 10mg
Chemical Name or Material Ozagrel hydrochloride
Assay Percent Range >98%
Molecular Formula C13H12N2O2·HCl
CAS 74003-18-2
Synonym (2E)-3-[4-(1H-Imidazol-1-ylmethyl)phenyl]-2-propenoic acid hydrochloride
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature