Learn More
Oxytetracycline hydrochloride, Thermo Scientific Chemicals
Impairs mitochondrial protein synthesis
$157.55 - $484.15
Chemical Identifiers
CAS | 2058-46-0 |
---|---|
Molecular Formula | C22H25ClN2O9 |
Molecular Weight (g/mol) | 496.897 |
MDL Number | MFCD00135815 |
InChI Key | UTAHTGPLLFLKGW-IFLJXUKPSA-N |
PubChem CID | 121235537 |
IUPAC Name | (4S,4aR,5S,5aR,6S,12aS)-4-(dimethylamino)-3,5,6,10,11,12a-hexahydroxy-6-methyl-1,12-dioxo-4,4a,5,5a-tetrahydrotetracene-2-carboxamide;hydrochloride |
SMILES | CC1(C2C(C3C(C(=C(C(=O)C3(C(=O)C2=C(C4=C1C=CC=C4O)O)O)C(=O)N)O)N(C)C)O)O.Cl |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAJ6248914
|
Thermo Scientific Chemicals
J6248914 |
25 g |
N/A
|
|
|||||
AAJ6248922
|
Thermo Scientific Chemicals
J6248922 |
100 g |
N/A
|
|
|||||
Description
Oxytetracycline is an antibiotic indicated for treatment of infections caused by Gram positive and Gram negative microorganisms such as Mycoplasma pneumoniae, Pasteurella pestis, Escherichia coli, Haemophilus influenzae, and Diplococcus pneumoniae. It is used in studies on the oxytetracycline-resistance gene (otrA). Oxytetracycline hydrochloride is used to study phagosome- lysosome (P-L) fusion in P388D1 cells1 and antibiotic susceptibilities of Mycoplasma bovis isolates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsOxytetracycline is an antibiotic indicated for treatment of infections caused by Gram positive and Gram negative microorganisms such as Mycoplasma pneumoniae, Pasteurella pestis, Escherichia coli, Haemophilus influenzae, and Diplococcus pneumoniae. It is used in studies on the oxytetracycline-resistance gene (otrA). Oxytetracycline hydrochloride is used to study phagosome- lysosome (P-L) fusion in P388D1 cells1 and antibiotic susceptibilities of Mycoplasma bovis isolates.
Solubility
Soluble in ethanol
Notes
Concentrated aqueous solutions at neutral pH will hydrolyze oxytetracycline and cause it to precipitate
Chemical Identifiers
2058-46-0 | |
496.897 | |
UTAHTGPLLFLKGW-IFLJXUKPSA-N | |
(4S,4aR,5S,5aR,6S,12aS)-4-(dimethylamino)-3,5,6,10,11,12a-hexahydroxy-6-methyl-1,12-dioxo-4,4a,5,5a-tetrahydrotetracene-2-carboxamide;hydrochloride |
C22H25ClN2O9 | |
MFCD00135815 | |
121235537 | |
CC1(C2C(C3C(C(=C(C(=O)C3(C(=O)C2=C(C4=C1C=CC=C4O)O)O)C(=O)N)O)N(C)C)O)O.Cl |
Specifications
Oxytetracycline Hydrochloride | |
2058-46-0 | |
C22H25ClN2O9 | |
3853107 | |
Soluble in ethanol | |
CC1(C2C(C3C(C(=C(C(=O)C3(C(=O)C2=C(C4=C1C=CC=C4O)O)O)C(=O)N)O)N(C)C)O)O.Cl | |
496.897 | |
496.9 | |
25 g | |
Oxytetracycline hydrochloride |
Crystalline Powder | |
180°C | |
MFCD00135815 | |
14,6976 | |
UTAHTGPLLFLKGW-IFLJXUKPSA-N | |
(4S,4aR,5S,5aR,6S,12aS)-4-(dimethylamino)-3,5,6,10,11,12a-hexahydroxy-6-methyl-1,12-dioxo-4,4a,5,5a-tetrahydrotetracene-2-carboxamide;hydrochloride | |
121235537 | |
Gray to Yellow | |
Air and light sensitive |
Safety and Handling
P201-P202-P264b-P280i-P281-P305+P351+P338-P308+P313-P501c
H319-H361d
EINECSNumber : 218-161-2
RTECSNumber : QI8225000
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only