missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Oxytetracycline hydrochloride, 50mg/mL in distilled water, sterile-filtered

$55.00 - $93.73

Chemical Identifiers

CAS 2058-46-0
Molecular Formula C22H25ClN2O9
Molecular Weight (g/mol) 496.897
MDL Number MFCD00135815
InChI Key UTAHTGPLLFLKGW-IFLJXUKPSA-N
PubChem CID 121235537
IUPAC Name (4S,4aR,5S,5aR,6S,12aS)-4-(dimethylamino)-3,5,6,10,11,12a-hexahydroxy-6-methyl-1,12-dioxo-4,4a,5,5a-tetrahydrotetracene-2-carboxamide;hydrochloride
SMILES CC1(C2C(C3C(C(=C(C(=O)C3(C(=O)C2=C(C4=C1C=CC=C4O)O)O)C(=O)N)O)N(C)C)O)O.Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ67398AC
View Documents
Alfa Aesar™
J67398AC
25mL Each for $55.00
 
AAJ67398AD
View Documents
Alfa Aesar™
J67398AD
50mL Each for $93.73
 
Specifications

Chemical Identifiers

2058-46-0
496.897
UTAHTGPLLFLKGW-IFLJXUKPSA-N
(4S,4aR,5S,5aR,6S,12aS)-4-(dimethylamino)-3,5,6,10,11,12a-hexahydroxy-6-methyl-1,12-dioxo-4,4a,5,5a-tetrahydrotetracene-2-carboxamide;hydrochloride
C22H25ClN2O9
MFCD00135815
121235537
CC1(C2C(C3C(C(=C(C(=O)C3(C(=O)C2=C(C4=C1C=CC=C4O)O)O)C(=O)N)O)N(C)C)O)O.Cl

Specifications

2058-46-0
MFCD00135815
CC1(C2C(C3C(C(=C(C(=O)C3(C(=O)C2=C(C4=C1C=CC=C4O)O)O)C(=O)N)O)N(C)C)O)O.Cl
496.897
Liquid
Light sensitive
C22H25ClN2O9
UTAHTGPLLFLKGW-IFLJXUKPSA-N
(4S,4aR,5S,5aR,6S,12aS)-4-(dimethylamino)-3,5,6,10,11,12a-hexahydroxy-6-methyl-1,12-dioxo-4,4a,5,5a-tetrahydrotetracene-2-carboxamide;hydrochloride
121235537
25mL
50mg/mL in distilled water, sterile-filtered
Safety and Handling

Safety and Handling

P201-P202-P281-P308+P313-P501c

H361d

EINECSNumber : 218-161-2

RTECSNumber : QI8225000

TSCA : Yes

storageNote1 : -30° to -10°C

Recommended Storage : Store at -20°C; Store under Argon