Learn More
Oxytetracycline, 96%, Thermo Scientific Chemicals
Oxytetracycline, CAS # 79-57-2, shows antibacterial, anti-inflammatory, and anti-proliferative activities. It can also inhibit mitochondrial protein synthesis.
Supplier: Thermo Scientific Chemicals J6242709
Description
Oxytetracycline is an anti-inflammatory, antibiotic, and anti-proliferative agent shown to inhibit mitochondrial protein synthesis. Mechanistic studies show that this compound reduces cytochrome c activity which eventually causes cell growth arrest. Additionally, Oxytetracycline has demonstrated the ability to reduce murine bronchial inflammation. The anti-inflammatory characteristics of this agent are due to its ability to cause a decrease in IL-4, IL-5, IL-13, immune cells, and the ovalbumin-specific IgE. Data suggests that Oxytetracycline is also an inhibitor of NF-κB activation via phosphorylation and degradation of IκB-α.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
- Oxytetracycline is a compound with broad-spectrum antibacterial activity. Furthermore, it also shows anti-inflammatory and anti-proliferative activities
- It can inhibit mitochondrial protein synthesis in both gram-positive and gram-negative bacteria
Application
- Oxytetracycline is active against several species including Mycoplasma pneumonia, Pasteurella pestis, Escherichia coli, Haemophilus influenza, and Diplococcus pneumonia
- In laboratory experiments, oxytetracycline can be used to evaluate the acquisition of oxytetracycline-resistance genes, which are associated with the development of antibiotic resistance
- It can reduce cytochrome c activity, causing cell growth arrest
- It has been shown to reduce murine bronchial inflammation
- Oxytetracycline exerts its anti-inflammatory activity by decreasing the levels of IL-4, IL-5, IL-13 immune cells, and the Ovalbumin-specific IgE
- Oxytetracycline can be used to inhibit NF-κB activation via phosphorylation and degradation of IκB-α
Specifications
Oxytetracycline | |
Powder | |
184°C to 185°C | |
MFCD00003700 | |
Sparingly soluble in aqueous solution at 0.6 mg/mL | |
O.O.[Na+].[Na+].CN(C)[C@H]1[C@@H]2[C@@H](O)[C@H]3[C-](C(=O)C4=C([O-])C=CC=C4[C@@]3(C)O)C(=O)[C@]2(O)C(=O)\C(=C(\N)O)C1=O | |
540.43 | |
460.43 | |
Tan to Yellow |
Oxytetracycline | |
79-57-2 | |
C22H26N2Na2O11 | |
14,6976 | |
OLRBUPVCLOPDOT-GETZKYKJSA-M | |
disodium (6aS,8Z,10S,10aR,11S,11aR,12S)-8-[amino(hydroxy)methylidene]-10-(dimethylamino)-6a,11,12-trihydroxy-12-methyl-5,6,7,9-tetraoxo-5,5a,6,6a,7,8,9,10,10a,11,11a,12-dodecahydrotetracen-5a-id-4-olate dihydrate | |
54675779 | |
≥98% | |
10 g |
Chemical Identifiers
79-57-2 | |
540.43 | |
OLRBUPVCLOPDOT-GETZKYKJSA-M | |
disodium (6aS,8Z,10S,10aR,11S,11aR,12S)-8-[amino(hydroxy)methylidene]-10-(dimethylamino)-6a,11,12-trihydroxy-12-methyl-5,6,7,9-tetraoxo-5,5a,6,6a,7,8,9,10,10a,11,11a,12-dodecahydrotetracen-5a-id-4-olate dihydrate |
C22H26N2Na2O11 | |
MFCD00003700 | |
54675779 | |
O.O.[Na+].[Na+].CN(C)[C@H]1[C@@H]2[C@@H](O)[C@H]3[C-](C(=O)C4=C([O-])C=CC=C4[C@@]3(C)O)C(=O)[C@]2(O)C(=O)\C(=C(\N)O)C1=O |
Safety and Handling
P201-P202-P261-P264b-P270-P271-P280i-P281-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P312-P330-P363-P501c
H302-H312-H319-H332-H360
EINECSNumber : 201-212-8
RTECSNumber : QI7875000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only