Please login to your online account to display your discounted pricing

Alfa Aesar™ Oxyntomodulin

For cellular and molecular biology applications

Specifications

Product Type Oxyntomodulin
Molecular Weight 4421.90
Storage Requirements -30° to -10°C
Sequence Glucagon 37
Format Solid
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66339EXD

 
alfa aesar™
J66339EXD
0.5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66339MCR

 
alfa aesar™
J66339MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type Oxyntomodulin
Molecular Weight 4421.90
Storage Requirements -30° to -10°C
Sequence Glucagon 37
Format Solid
Regulatory Status RUO
Purification Method HPLC