missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Oxonic acid potassium salt, Thermo Scientific Chemicals

Inhibitor of uricase

Supplier:  Thermo Scientific Chemicals J6040022

 View more versions of this product

Catalog No. AAJ6040022



Description

Description

Oxonic acid potassium salt is an inhibitor of uricase. This product has been used for the inhibition of 5-fluorouracil-induced gastrointestinal toxicity without the loss of its antitumor activity in rats. It has also been used to induce hyperuricemia in rats; as it inhibits uric acid metabolism.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Oxonic acid potassium salt is an inhibitor of uricase. This product has been used for the inhibition of 5-fluorouracil-induced gastrointestinal toxicity without the loss of its antitumor activity in rats. It has also been used to induce hyperuricemia in rats; as it inhibits uric acid metabolism.

Solubility
Soluble in hot water.

Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agents.
Specifications

Specifications

Oxonic acid potassium salt
98%
2207-75-2
100 g
MFCD00010565
potassium oxonate, oteracil potassium, oxonic acid potassium salt, allantoxanic acid potassium salt, potassium azaorotate, oxonic acid, potassium salt, oxonate, potassium, oxonate, allantoxanic acid
IAPCTXZQXAVYNG-UHFFFAOYSA-M
potassium;4,6-dioxo-1H-1,3,5-triazine-2-carboxylate
2723920
195.18
300°C
White
Powder
C4H2KN3O4
14,6947
Soluble in hot water.
[K+].[O-]C(=O)C1=NC(=O)NC(=O)N1
195.18
CHEBI:80230

Chemical Identifiers

2207-75-2
195.18
IAPCTXZQXAVYNG-UHFFFAOYSA-M
2723920
[K+].[O-]C(=O)C1=NC(=O)NC(=O)N1
C4H2KN3O4
MFCD00010565
potassium oxonate, oteracil potassium, oxonic acid potassium salt, allantoxanic acid potassium salt, potassium azaorotate, oxonic acid, potassium salt, oxonate, potassium, oxonate, allantoxanic acid
CHEBI:80230
Safety and Handling

Safety and Handling

EINECSNumber : 218-627-5

RTECSNumber : RR4580000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only