Thermo Scientific™ Oxoid™ VCNT Selective Supplement
The Oxoid VCNT Selective Supplement is used for the preparation of Thayer Martin Medium for the isolation of pathogenic Neisseria spp.
$278.47 - $598.48
Specifications
Description | VCNT Selective Supplement |
---|---|
Format | Freeze Dried |
Packaging | 10/Pk. |
Quantity | 10 x Freeze Dried Vials |
Description
Oxoid VCNT Selective Supplement is used for the preparation of Thayer Martin Medium for the isolation of pathogenic Neisseria spp.
- Add to GC Agar Base, Part No. CM0367B
Specifications
VCNT Selective Supplement | |
Freeze Dried | |
10/Pk. | |
10 x Freeze Dried Vials |