Thermo Scientific™ Oxoid™ UVM1 Listeria Primary Selective Enrichment Supplement
The Thermo Scientific™ Oxoid Listeria Primary Selective Enrichment Supplement, UVM1 is a selective method for enrichment of Listeria spp.
Supplier: Thermo Scientific™ SR0142E
Description
Thermo Scientific™ Oxoid Listeria Primary Selective Enrichment Supplement, UVM1 is used in a two-step selective enrichment (USDA-FSIS) method for the enrichment of Listeria spp.
Add to Listeria Enrichment Broth Base (UVM formulation), Part No. CM0863BSpecifications
Listeria | |
Freeze-dried | |
10/Pk. |
Listeria Primary Selective Enrichment Supplement, UVM1 | |
Selective Supplement | |
Each vial for 500mL medium |