Thermo Scientific™ Tryptone Soy Broth (Soybean Casein Digest Medium EP/USP/BP/JP) (Dehydrated)
Grow a wide variety of microorganisms with this general purpose medium containing two peptones.
$194.53 - $1835.81
Specifications
Description Tryptone Soy Broth (Soybean Casein Digest Medium USP)
Form Powder
Product Type Dehydrated Culture Media
View More Specs
Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
OXCM0129R
View Documents Promotion Details
Thermo Scientific™
CM0129R
2.5 kg
Each for $992.37
OXCM0129B
View Documents Promotion Details
Thermo Scientific™
CM0129B
500 g
Each for $194.53
OXCM0129T
View Documents Promotion Details
Thermo Scientific™
CM0129T
5 kg
Each for $1,835.81
Description
Description
Grow a wide variety of microorganisms with Thermo Scientific™ Oxoid™ Tryptone Soya Broth EP/USP/BP/JP (Dehydrated). The general purpose medium, containing two peptones, is suitable for the cultivation of both aerobes and anaerobes. It is specified in International Pharmacopoeia1,2,3,4 and food testing methods for the growth of a wide variety of organisms.
Use Tryptone Soya Broth to support the luxuriant growth of many fastidious organisms, without the addition of serum, while conforming to the formulations detailed in various international pharmacopoeias.
  • Suitable for tube dilution method of antibiotic susceptibility testing
  • Recommended in the USP for the recovery of organisms after sterilization processes4
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand
Specifications
Specifications
Tryptone Soy Broth (Soybean Casein Digest Medium USP)
Powder
Dehydrated Culture Media
SDS
missing translation for 'documents'
missing translation for 'documents'
For Laboratory Use Only