Thermo Scientific™ Oxoid™ Oxford Listeria Selective Supplement
The Oxoid Listeria Selective Supplement (Oxford) is used for the detection of Listeria monocytogenes.
Manufacturer:  Thermo Scientific™ SR0206E
 View more versions of this product
Catalog No. OXSR0206E
Description
Description
Oxoid Listeria Selective Supplement (Oxford) is used for the detection of Listeria monocytogenes.
  • Add to Listeria Selective Agar Base, Part No. CM0856B
Specifications
Specifications
Freeze Dried
Listeria Selective Supplement
Listeria
10/Pk.
Supplement
Selective Supplement
Each vial for 500mL medium