Thermo Scientific™ Oxoid™ Oxford Listeria Selective Supplement
The Oxoid Listeria Selective Supplement (Oxford) is used for the detection of Listeria monocytogenes.
Manufacturer:  Thermo Scientific™ SR0206B
 View more versions of this product
Catalog No. OXSR0206B
Description
Description
Oxoid Listeria Selective Supplement (Oxford) is used for the detection of Listeria monocytogenes.
  • Add to Listeria Selective Agar Base, Part No. CM0856B
Specifications
Specifications
Mod Lister(oxf)sup For 2.5L 10Pk.
Selective Supplement
Listeria
For 2.5L medium