missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Oxoid™ ONE Broth-Listeria Supplement

The Oxoid ONE Broth-Listeria is for the selective enrichment broth for Listeria species from food samples in 24 hours.

$313.44 - $313.44

Specifications

Species Listeria
Description ONE Broth-Listeria Supplement
Form Freeze-dried
Product Type Microbiology Media
Quantity 10/Pk.
View More Specs

Products 1
Catalog Number Mfr. No. Yield Price Quantity  
Catalog Number Mfr. No. Yield Price Quantity  
OXSR0234E
View Documents Promotion Details
Thermo Scientific™
SR0234E
Each vial for 225mL medium
Each for $313.44
 
Description

Description

The Oxoid ONE Broth-Listeria is for the selective enrichment broth for Listeria species from food samples in 24 hours.

Add to ONE Broth-Salmonella, Part No. CM1091B.

ONE Broth - Listeria: Typical Formulation

  • Peptone: 26g/L; Carbohydrate mix: 6g/L; Salt mix: 10g/L with a final pH of 7.4 ±0.2 at 25°C

Appearance:

  • Dehydrated medium: straw coloured, free-flowing powder
  • Selective supplement: orange/yellow, freeze-dried pellet
  • Prepared medium: dark straw coloured, clear liquid

Note:

Formulation Directions

  • Suspend 22g of ONE Broth-Listeria Base in 500mL (44g/L) of distilled water
  • Mix well and sterilize by autoclaving at 121°C for 15 minutes
  • Cool the medium to below 50°C and aseptically add the contents of one vial of ONE Broth-Listeria Selective Supplement (Mfr. No. SR0234) reconstituted as directed

Testing Protocol

  • Add 25g of food sample to 225mL of ONE Broth-Listeria and stomach for a minimum of 30 seconds to mix the sample
  • Incubate the broth without agitation at 30°C for 24±2 hours
  • Gently agitate the bag then, using a microbiological loop, remove 10μL and inoculate onto a suitable agar medium e.g. Brilliance Listeria Agar (Mfr. No. CM1080, SR0222, and SR0228) and incubate appropriately (37°C for 24-48 hours)

  • Confirm presumptive colonies on the agar plates as Listeria monocytogenes or Listeria spp. by appropriate methods e.g. Gram-stain, catalase, oxidase, Oxoid Listeria Latex Test Kit (Mfr. No. DR1126)
  • Oxoid O.B.I.S. mono (Mfr. No. ID0600), Microbact Listeria 12L (Mfr. No. MB1128)

Specifications

Specifications

Listeria
Freeze-dried
10/Pk.
ONE Broth-Listeria Supplement
Microbiology Media
SDS
missing translation for 'documents'

missing translation for 'documents'

For Laboratory Use Only