Thermo Scientific™ Oxoid™ Novobiocin Antimicrobial Susceptibility Disks
Determine the susceptibility of microorganisms using manual AST methods and a full range of high quality, easy-to-use antimicrobial susceptibility testing disks.
$54.79 - $54.79
Specifications
Description | Oxoid Novobiocin Antimicrobial Susceptibility Disks |
---|---|
Quantity | 50 Disks/Cartridge |
Packaging | 50 Disks/Cartridge |
Catalog Number | Mfr. No. | Description | Concentration | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Description | Concentration | Price | Quantity | ||||
OXCT0038B
|
Thermo Scientific™
CT0038B |
Oxoid Novobiocin Antimicrobial Susceptibility Disks | 30μg |
Each for $54.79
|
|
Description
Manually determine the antibiotic susceptibility of microorganisms using Antimicrobial Susceptibility Testing (AST) methods in conjunction with reliable, easy-to-use Thermo Scientific™ Oxoid™ Novobiocin Antimicrobial Susceptibility Disks.
Ease of use is provided by shatterproof spring-loaded cartridges, easy-to-read labeling and end-of-cartridge indicators.Disk diffusion not only categorizes resistant, intermediate and sensitive organisms through quantitative results, but also provides a visual indication of the following:
- Inoculum level
- Presence of contamination
- Resistant mutants
- Beta-lactamase activity
- Antagonism and synergy between antibiotics
Each cartridge is individually sealed with a desiccant to maintain a moisture level of <2%, ensuring long term stability.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need, and the quality results your laboratory depends on.
Specifications
Oxoid Novobiocin Antimicrobial Susceptibility Disks | |
50 Disks/Cartridge | |
50 Disks/Cartridge |