Thermo Scientific™ Oxoid™ Listeria Selective Supplement
The Oxoid Listeria Selective Supplement (Oxford) is used for the detection of Listeria monocytogenes.
$261.35 - $1,041.31
Specifications
Description | Listeria Selective Supplement, Oxford |
---|---|
Format | Freeze Dried |
Packaging | 10/Pk. |
Quantity | 10 x Freeze Dried Vials |
Description
Oxoid Listeria Selective Supplement (Oxford) is used for the detection of Listeria monocytogenes.
Add to Listeria Selective Agar Base, Part No. CM0856BSpecifications
Listeria Selective Supplement, Oxford | |
Freeze Dried | |
10/Pk. | |
10 x Freeze Dried Vials |