Thermo Scientific™ Oxoid™ Listeria Selective Supplement
The Oxoid Listeria Selective Supplement (Oxford) is used for the detection of Listeria monocytogenes.
Supplier: Thermo Scientific™ SR0140B
Description
Oxoid Listeria Selective Supplement (Oxford) is used for the detection of Listeria monocytogenes.
Add to Listeria Selective Agar Base, Part No. CM0856BSpecifications
Listeria | |
Freeze-dried | |
10/Pk. |
Listeria Selective Supplement, Oxford | |
Selective Supplement | |
Each vial for 2.5L medium |