Thermo Scientific™ Oxoid™ Legionella MWY Selective Supplement
Oxoid Legionella MWY Selective Supplement is used for the isolation of Legionella pneumophila from potable water samples.
$351.44 - $1,210.52
Specifications
Description | Legionella MWY Selective Supplement |
---|---|
Format | Freeze Dried |
Packaging | 10/Pk. |
Quantity | 10 x Freeze Dried Vials |
Description
Oxoid Legionella MWY Selective Supplement is used for the isolation of Legionella pneumophila from potable water samples.
Add to Legionella Agar Base, Part No. CM1203BSpecifications
Legionella MWY Selective Supplement | |
Freeze Dried | |
10/Pk. | |
10 x Freeze Dried Vials |