missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Thermo Scientific™ Oxoid™ Half Fraser Supplement
The Oxoid Half Fraser Supplement is used for the enrichment of Listeria species.
$322.84 - $890.80
Specifications
Species | Listeria |
---|---|
Description | Half Fraser Supplement |
Form | Freeze-dried |
Product Type | Microbiology Media |
Description
Oxoid Half Fraser Supplement is used for the enrichment of Listeria species.
- Add to Fraser Broth Base, Part No. CM0895B
Specifications
Listeria | |
Half Fraser Supplement | |
Freeze-dried | |
Microbiology Media |