missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Oxoid™ Dehydrated Culture Media

$185.70 - $194.50

Specifications

Format Dehydrated
Storage Requirements 15° to 30°C
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Media Type Product Type Price Quantity  
Catalog Number Mfr. No. Media Type Product Type Price Quantity  
OXCM0149B Thermo Scientific™
CM0149B
Reinforced Clostridial Medium Reinforced Clostridial medium Each for $185.70
OXCM0151B Thermo Scientific™
CM0151B
Reinforced Clostridial Agar Reinforced Clostridial Agar Each for $194.50
Specifications

Description

Oxoid is a world leading microbiology brand which has been associated with quality culture media for over 50 years. The wide range of Oxoid media products is trusted by clinical, industrial and academic microbiology laboratories to provide the excellent performance they need for their microbiological testing.

Description & Specifications

Specifications

Dehydrated
15° to 30°C