Thermo Scientific™ Oxoid™ Anaerobic Low Temperature Catalyst
Oxoid Anaerobic Low Temperature Catalyst is a safety catalyst designed to stay cooler than traditional catalysts.
Supplier: Thermo Scientific™ BR0042A
Description
Oxoid Anaerobic Low Temperature Catalyst is a safety catalyst designed to stay cooler than traditional catalysts.
- To be changed every 30 cycles
Specifications
Anaerobic Low Temperature Catalyst | |
Each |