missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Oxindole, 97+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1876406
Description
Oxindole is considered as indole analogue, which shows pharmacological activity.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsOxindole is considered as indole analogue, which shows pharmacological activity.
Solubility
Soluble in methanol, ethanol, dimethyl sulfoxide and ether. Insoluble in water.
Notes
Incompatible with strong oxidizing agents.
Specifications
Oxindole | |
124°C to 128°C | |
C8H7NO | |
5 g | |
oxindole, indolin-2-one, 2-oxindole, 2-indolinone, 1,3-dihydro-2h-indol-2-one, oxindol, 2-oxoindoline, 2-oxindoline, indol-2 3h-one, 2h-indol-2-one, 1,3-dihydro | |
JYGFTBXVXVMTGB-UHFFFAOYSA-N | |
1,3-dihydroindol-2-one | |
321710 | |
133.15 |
59-48-3 | |
194°C to 196°C (17 mmHg) | |
MFCD00005711 | |
114692 | |
Soluble in methanol,ethanol,dimethyl sulfoxide and ether. Insoluble in water. | |
C1C2=CC=CC=C2NC1=O | |
133.15 | |
CHEBI:31697 | |
≥97% |
Chemical Identifiers
59-48-3 | |
133.15 | |
JYGFTBXVXVMTGB-UHFFFAOYSA-N | |
321710 | |
1,3-dihydroindol-2-one |
C8H7NO | |
MFCD00005711 | |
oxindole, indolin-2-one, 2-oxindole, 2-indolinone, 1,3-dihydro-2h-indol-2-one, oxindol, 2-oxoindoline, 2-oxindoline, indol-2 3h-one, 2h-indol-2-one, 1,3-dihydro | |
CHEBI:31697 | |
C1C2=CC=CC=C2NC1=O |
Safety and Handling
GHS H Statement
H302
Harmful if swallowed.
P264b-P270-P301+P312-P330-P501c
H302
EINECSNumber : 200-429-5
RTECSNumber : NM2080500
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only