Please login to your online account to display your discounted pricing

Alfa Aesar™ Oxidized ACTH (1-10), Certified Mass Spec Grade


For cellular and molecular biology applications

Manufacturer: alfa aesar™  J67553LPL

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. AAJ67553LPL

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Quantity 1 vial, 500 pmol
Format Lyophilized powder
Sequence SYS-M(O)-EHFRWG
Regulatory Status RUO
Storage Requirements -30° to -10°C
Product Type Oxidized ACTH (1-10), Certified Mass Spec Grade
Molecular Weight 1315.40
Purification Method HPLC