missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Oxibendazole 98.0+%, TCI America™

Manufacturer:  TCI America O0499500MG

 View more versions of this product

Catalog No. O0499500MG



Specifications

Specifications

Oxibendazole
230°C
20559-55-1
MFCD00133728
RAOCRURYZCVHMG-UHFFFAOYSA-N
methyl N-(6-propoxy-1H-benzimidazol-2-yl)carbamate
4622
Crystal-Powder at 20°C
Undesignated
500mg
C12H15N3O3
Methyl (5-Propoxy-1H-benzo[d]imidazol-2-yl)carbamate, (5-Propoxy-1H-benzimidazol-2-yl)carbamic Acid Methyl Ester
CCCOC1=CC2=C(C=C1)N=C(N2)NC(=O)OC
249.27
249.27
≥98.0% (HPLC,N)

Chemical Identifiers

20559-55-1
249.27
RAOCRURYZCVHMG-UHFFFAOYSA-N
4622
CCCOC1=CC2=C(C=C1)N=C(N2)NC(=O)OC
C12H15N3O3
MFCD00133728
Methyl (5-Propoxy-1H-benzo[d]imidazol-2-yl)carbamate, (5-Propoxy-1H-benzimidazol-2-yl)carbamic Acid Methyl Ester
methyl N-(6-propoxy-1H-benzimidazol-2-yl)carbamate
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator

Documents