Learn More
Oxetan-3-ol, 95%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 435840010
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Oxetan-3-ol | |
1.1250g/mL | |
87°C | |
94% min. (GC) | |
C3H6O2 | |
1g | |
3-hydroxyoxetane, 3-oxetanol, 1,3-epoxy-2-propanol, oxetane-3-ol, 3-hydroxy-oxetane, 3-hydroxy oxetane, pubchem17367, ksc380k3t, 1,3-epoxypropan-2-ol | |
OC1COC1 | |
74.08 | |
74.08 |
7748-36-9 | |
153°C | |
Authentic | |
Glass bottle | |
MFCD09056790 | |
1.125 | |
QMLWSAXEQSBAAQ-UHFFFAOYSA-N | |
oxetan-3-ol | |
9942117 | |
95% |
Chemical Identifiers
7748-36-9 | |
74.08 | |
QMLWSAXEQSBAAQ-UHFFFAOYSA-N | |
9942117 | |
OC1COC1 |
C3H6O2 | |
MFCD09056790 | |
3-hydroxyoxetane, 3-oxetanol, 1,3-epoxy-2-propanol, oxetane-3-ol, 3-hydroxy-oxetane, 3-hydroxy oxetane, pubchem17367, ksc380k3t, 1,3-epoxypropan-2-ol | |
oxetan-3-ol |
Safety and Handling
GHS H Statement
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Danger
RUO â Research Use Only