missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Oxetan-3-ol, 95%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC435840050
Specifications
Oxetan-3-ol | |
87°C | |
Glass bottle | |
153°C | |
5g | |
94% min. (GC) | |
1,3-epoxy-2-propanol, 1,3-epoxypropan-2-ol, 3-hydroxy oxetane, 3-hydroxy-oxetane, 3-hydroxyoxetane, 3-oxetanol, ksc380k3t, oxetane-3-ol, pubchem17367 | |
C1C(CO1)O | |
74.08 | |
74.08 | |
95% |
1.1250g/mL | |
Authentic | |
1.125 | |
Colorless | |
7748-36-9 | |
C3H6O2 | |
QMLWSAXEQSBAAQ-UHFFFAOYSA-N | |
oxetan-3-ol | |
9942117 | |
Liquid |
Chemical Identifiers
7748-36-9 | |
74.08 | |
1,3-epoxy-2-propanol, 1,3-epoxypropan-2-ol, 3-hydroxy oxetane, 3-hydroxy-oxetane, 3-hydroxyoxetane, 3-oxetanol, ksc380k3t, oxetane-3-ol, pubchem17367 | |
oxetan-3-ol |
C3H6O2 | |
QMLWSAXEQSBAAQ-UHFFFAOYSA-N | |
9942117 | |
C1C(CO1)O |
Safety and Handling
GHS H Statement
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Danger