Learn More
Oxazole-5-carboxylic acid, 98+%, Thermo Scientific Chemicals
$109.41 - $325.45
Chemical Identifiers
CAS | 118994-90-4 |
---|---|
Molecular Formula | C4H3NO3 |
Molecular Weight (g/mol) | 113.072 |
MDL Number | MFCD04114931 |
InChI Key | QCGMEWVZBGQOFN-UHFFFAOYSA-N |
Synonym | oxazole-5-carboxylic acid, 5-oxazolecarboxylic acid, 5-oxazolecarboxylicacid, 5-carboxy-1,3-oxazole, oxazole-5-carboxylicacid, 5-carboxyoxazole, acmc-209a0v, ksc173g8b, 1,3-oxazole-5-carboxylic acid |
PubChem CID | 16340557 |
IUPAC Name | 1,3-oxazole-5-carboxylic acid |
SMILES | C1=C(OC=N1)C(=O)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH3017903
|
Thermo Scientific Chemicals
H3017903 |
1 g |
Each for $109.41
|
|
|||||
AAH3017906
|
Thermo Scientific Chemicals
H3017906 |
5 g |
Each for $325.45
|
|
|||||
Description
Decarboxylative cross-coupling of thiazole and oxazole-5-carboxylic acids with aryl halides is reported.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsDecarboxylative cross-coupling of thiazole and oxazole-5-carboxylic acids with aryl halides is reported.
Solubility
Soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Chemical Identifiers
118994-90-4 | |
113.072 | |
QCGMEWVZBGQOFN-UHFFFAOYSA-N | |
16340557 | |
C1=C(OC=N1)C(=O)O |
C4H3NO3 | |
MFCD04114931 | |
oxazole-5-carboxylic acid, 5-oxazolecarboxylic acid, 5-oxazolecarboxylicacid, 5-carboxy-1,3-oxazole, oxazole-5-carboxylicacid, 5-carboxyoxazole, acmc-209a0v, ksc173g8b, 1,3-oxazole-5-carboxylic acid | |
1,3-oxazole-5-carboxylic acid |
Specifications
118994-90-4 | |
C4H3NO3 | |
1 g | |
Soluble in water. | |
C1=C(OC=N1)C(=O)O | |
113.072 | |
113.07 | |
Oxazole-5-carboxylic acid |
∽245°C (decomposition) | |
MFCD04114931 | |
oxazole-5-carboxylic acid, 5-oxazolecarboxylic acid, 5-oxazolecarboxylicacid, 5-carboxy-1,3-oxazole, oxazole-5-carboxylicacid, 5-carboxyoxazole, acmc-209a0v, ksc173g8b, 1,3-oxazole-5-carboxylic acid | |
QCGMEWVZBGQOFN-UHFFFAOYSA-N | |
1,3-oxazole-5-carboxylic acid | |
16340557 | |
≥98% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only