Please login to your online account to display your discounted pricing

Alfa Aesar™ Oxanilide, 97%

$117.61 - $117.61

Chemical Identifiers

Chemical Name or Material Oxanilide
CAS 620-81-5
Molecular Formula C14H12N2O2
Formula Weight 240.26
MDL Number MFCD00041758
Beilstein 1645104
Synonym N,N'-Diphenyloxamide; Oxalic acid dianilide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0736914

 
alfa aesar™
L0736914
25g Each for $117.61
Description & Specifications

Specifications

Formula Weight 240.26
Chemical Name or Material Oxanilide
Molecular Formula C14H12N2O2
TSCA TSCA
Assay 97%
MDL Number MFCD00041758
CAS 620-81-5
Beilstein 1645104
Synonym N,N'-Diphenyloxamide; Oxalic acid dianilide
RTECS Number KH8368000
Melting Point 252° to 254°C
Quantity 25g