missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Oxamide, 98+%

Manufacturer:  Alfa Aesar™ A1157036

 View more versions of this product

Catalog No. AAA1157036


 

Description

Description

Applications
Oxamide is used as a stabilizer for nitrocellulose preparations. It also finds use in APCP rocket motors as a high performance burn rate suppressant.

Solubility
Soluble in ethanol, slightly soluble in water and insoluble in diethyl ether.

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
Specifications

Specifications

Oxamide
≥98%
MFCD00008007
14,6918
YIKSCQDJHCMVMK-UHFFFAOYSA-N
oxamide
10113
88.07
Odorless
471-46-5
C2H4N2O2
1743262
Oxalic diamide
C(=O)(C(=O)N)N
88.066
CHEBI:48248
~350°C (decomposition)
500g

Chemical Identifiers

471-46-5
88.066
YIKSCQDJHCMVMK-UHFFFAOYSA-N
10113
C2H4N2O2
MFCD00008007
Oxalic diamide
CHEBI:48248
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : RO4900000

TSCA : TSCA

SDS Documents