missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Oxamic Hydrazide 99%, ACROS Organics™

Manufacturer:  Fisher Scientific Acros 129640250

 View more versions of this product

Catalog No. AC129640250

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

≥98.5%
Powder
White
15, 8584
Plastic bottle
99%
99%
MFCD00008004
C2H5N3O2
103.08
25g
02, 559
Semioxamazide
Authentic
Oxamic hydrazide
515-96-8
H2NCOCONHNH2

Chemical Identifiers

515-96-8
C2H5N3O2
103.08
Semioxamazide
≥98.5%
99%
H2NCOCONHNH2
MFCD00008004
Oxamic hydrazide, 99%

Safety and Handling

SDS & Safety Documents

EINECSNumber : 208-213-2

RTECSNumber : AF2620000