missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Oxamic acid, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals B2034414

 View more versions of this product

Catalog No. AAB2034414



Description

Description

Oxamic acid could also inhibit lactic dehydrogenase in the intact Ehrlich ascites tumor cell.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Oxamic acid could also inhibit lactic dehydrogenase in the intact Ehrlich ascites tumor cell.

Solubility
Soluble in water 108 mg/mL.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
Specifications

Specifications

Oxamic acid
∼205°C (decomposition)
MFCD00008006
1743294
2-amino-2-oxoacetic acid, oxamate, acetic acid, aminooxo, oxalamic acid, oxamidic acid, glycine, 2-oxo, oxalic acid monoamide, amino oxo acetic acid, aminooxoacetic acid, oxamate repellent
SOWBFZRMHSNYGE-UHFFFAOYSA-N
oxamic acid
974
89.05
471-47-6
C2H3NO3
25 g
14,6917
Soluble in water 108mg/mL.
C(=O)(C(=O)O)N
89.05
CHEBI:18058
98%

Chemical Identifiers

471-47-6
89.05
SOWBFZRMHSNYGE-UHFFFAOYSA-N
974
oxamic acid
C2H3NO3
MFCD00008006
2-amino-2-oxoacetic acid, oxamate, acetic acid, aminooxo, oxalamic acid, oxamidic acid, glycine, 2-oxo, oxalic acid monoamide, amino oxo acetic acid, aminooxoacetic acid, oxamate repellent
CHEBI:18058
C(=O)(C(=O)O)N
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 207-443-0

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only