missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Oxalyl chloride, 98%, Alfa Aesar

Manufacturer:  Alfa Aesar A1801218*

 View more versions of this product

Catalog No. AAA1801218


 

Description

Description

Applications
Oxalyl chloride is mainly used as a catalyst in the oxidation of alcohols to the corresponding aldehydes and ketones. It is actively used for the synthesis of acid chlorides from acids. In the Fridel-Crafts acylation reaction, it reacts with aromatic compounds to get the corresponding acyl choride using aluminum chloride as catalyst. Further, it is utilized to prepare dioxane tetrketone, an oxide of carbon.

Solubility
Oxalyl chloride Slightly miscible with water.Miscible with ether, benzene and chloroform.

Notes
Moisture sensitive. Incompatible with bases, alcohols, water, amines and metals.
Specifications

Specifications

Oxalyl chloride
1.455
0.98
MFCD00000704
1361988
CTSLXHKWHWQRSH-UHFFFAOYSA-N
oxalyl dichloride
65578
Liquid
79-37-8
98%
C2Cl2O2
UN2927
14,6914
C(=O)(C(=O)Cl)Cl
126.92
126.93
50g

Chemical Identifiers

79-37-8
126.92
CTSLXHKWHWQRSH-UHFFFAOYSA-N
C2Cl2O2
MFCD00000704
65578
Safety and Handling

Safety and Handling

GHS H Statement
H331-H314-H318-H302

GHS P Statement
P280-P303+P361+P353-P305+P351+P338-P310-P402+P404

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC LIQUIDS, CORROSIVE, ORGANIC, N.O.S.

RTECSNumber : KI2950000

TSCA : TSCA

storageNote1 : Moisture Sensitive

SDS Documents