Learn More
Oxalyl chloride, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1801222
Description
Oxalyl chloride is mainly used as a catalyst in the oxidation of alcohols to the corresponding aldehydes and ketones. It is actively used for the synthesis of acid chlorides from acids. In the Fridel-Crafts acylation reaction, it reacts with aromatic compounds to get the corresponding acyl choride using aluminum chloride as catalyst. Further, it is utilized to prepare dioxane tetrketone, an oxide of carbon.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsOxalyl chloride is mainly used as a catalyst in the oxidation of alcohols to the corresponding aldehydes and ketones. It is actively used for the synthesis of acid chlorides from acids. In the Fridel-Crafts acylation reaction, it reacts with aromatic compounds to get the corresponding acyl choride using aluminum chloride as catalyst. Further, it is utilized to prepare dioxane tetrketone, an oxide of carbon.
Solubility
Oxalyl chloride Slightly miscible with water.Miscible with ether, benzene and chloroform.
Notes
Moisture sensitive. Incompatible with bases, alcohols, water, amines and metals.
Specifications
Oxalyl chloride | |
-12°C | |
63°C to 64°C | |
Acrid | |
1.429 | |
100 g | |
1361988 | |
14,6914 | |
Oxalyl chloride Slightly miscible with water.Miscible with ether,benzene and chloroform. | |
C(=O)(C(=O)Cl)Cl | |
126.92 | |
126.93 | |
Liquid |
79-37-8 | |
1.455 | |
0.98 | |
C2Cl2O2 | |
MFCD00000704 | |
UN2927 | |
Moisture sensitive | |
oxalyl chloride, ethanedioyl dichloride, oxalic dichloride, oxaloyl chloride, oxalic acid dichloride, oxalic acid chloride, ethanedioyl chloride, oxalylchloride, unii-r4y96317dw, cocl 2 | |
CTSLXHKWHWQRSH-UHFFFAOYSA-N | |
oxalyl dichloride | |
65578 | |
98% |
Chemical Identifiers
79-37-8 | |
126.92 | |
CTSLXHKWHWQRSH-UHFFFAOYSA-N | |
65578 | |
C(=O)(C(=O)Cl)Cl |
C2Cl2O2 | |
MFCD00000704 | |
oxalyl chloride, ethanedioyl dichloride, oxalic dichloride, oxaloyl chloride, oxalic acid dichloride, oxalic acid chloride, ethanedioyl chloride, oxalylchloride, unii-r4y96317dw, cocl 2 | |
oxalyl dichloride |
Safety and Handling
GHS H Statement
H331-H314-H318-H302
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
P223-P231+P232-P260-P264b-P270-P271-P280-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P335+P334-P363-P370+P378q-P501c
H260-H301+H331-H314
DOTInformation : Hazard Class: 4.3; Packaging Group: I
EINECSNumber : 201-200-2
RTECSNumber : KI2950000
TSCA : Yes
storageNote1 : Moisture Sensitive
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only